Please login to your online account to display your discounted pricing

2-Hydroxy-5-nitropyridine 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-5-nitropyridine
Name Note 99+%
CAS 5418-51-9
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H4N2O3
Linear Formula HO(C5H3N)NO2
Formula Weight 140.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121680100

  SDS
fisher scientific acros
121680100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC121681000

  SDS
thermo fisher scientific
121681000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 140.1
Quantity 10g
Color Light yellow to beige
Beilstein 21, I, 202
EINECS Number 226-525-7
TSCA TSCA
Melting Point 188° to 191°C
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 2-Hydroxy-5-nitropyridine
Packaging Glass bottle
Name Note 99+%
Molecular Formula C5H4N2O3
Synonym 5-Nitro-2-pyridinol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula HO(C5H3N)NO2
MDL Number MFCD00006276
CAS 5418-51-9
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.