Please login to your online account to display your discounted pricing

2-Hydroxy-5-nitrobenzaldehyde 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC416181000

 View more versions of this product

Catalog No. AC416181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystals
Formula Weight 167.12
Assay ≥98.0%
Quantity 100g
Color Yellow to orange to brown
TSCA TSCA
RTECS Number CU6675000
EINECS Number 202-587-0
Melting Point 124° to 128°C
CAS Min % 98.0
CAS Max % 100.0
Synonym 5-Nitrosalicaldehyde
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
CAS 97-51-8
Chemical Name or Material 2-Hydroxy-5-nitrobenzaldehyde
Name Note 99%
Assay Percent Range 98+%
Molecular Formula C7H5NO4
MDL Number MFCD00007337

Specifications

Chemical Name or Material 2-Hydroxy-5-nitrobenzaldehyde
Name Note 99%
CAS 97-51-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H5NO4
Formula Weight 167.12
MDL Number MFCD00007337
Synonym 5-Nitrosalicaldehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents