missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-5-nitrobenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1026406

 View more versions of this product

Catalog No. AAA1026406



Description

Description

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Hydroxy-5-nitrobenzaldehyde
5g
98%
MFCD00007337
5-Nitrosalicylaldehyde; 5-Nitrosalicylaldehyde
C1=CC(=C(C=C1[N+](=O)[O-])C=O)O
167.12
167.12
123°C to 127°C
97-51-8
C7H5NO4
512565
IHFRMUGEILMHNU-UHFFFAOYSA-N
2-hydroxy-5-nitrobenzaldehyde
66808

Chemical Identifiers

97-51-8
167.12
IHFRMUGEILMHNU-UHFFFAOYSA-N
66808
C1=CC(=C(C=C1[N+](=O)[O-])C=O)O
C7H5NO4
MFCD00007337
5-Nitrosalicylaldehyde; 5-Nitrosalicylaldehyde
2-hydroxy-5-nitrobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : CU6675000

TSCA : TSCA

SDS Documents