missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-5-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1026414

 View more versions of this product

Catalog No. AAA1026414



Description

Description

2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Hydroxy-5-nitrobenzaldehyde
123°C to 127°C
MFCD00007337
512565
5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
IHFRMUGEILMHNU-UHFFFAOYSA-N
2-hydroxy-5-nitrobenzaldehyde
66808
98%
97-51-8
C7H5NO4
25 g
Air and light sensitive
Soluble in water (partly).
OC1=CC=C(C=C1C=O)[N+]([O-])=O
167.12
167.12

Chemical Identifiers

97-51-8
167.12
IHFRMUGEILMHNU-UHFFFAOYSA-N
66808
OC1=CC=C(C=C1C=O)[N+]([O-])=O
C7H5NO4
MFCD00007337
5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
2-hydroxy-5-nitrobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-587-0

RTECSNumber : CU6675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only