missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2′-Hydroxy-5′-methylpropiophenone, TCI America™

$62.64 - $212.36

Chemical Identifiers

CAS 938-45-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD01098830
InChI Key CXZJBPYDVCLMFX-UHFFFAOYSA-N
Synonym 2-Propionyl-p-cresol
PubChem CID 70311
IUPAC Name 1-(2-hydroxy-5-methylphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=CC(=C1)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14941G View Documents TCI America
H14941G
1g Each for $62.64
 
H14945G View Documents TCI America
H14945G
5g Each for $212.36
 
Specifications

Chemical Identifiers

938-45-4
164.204
CXZJBPYDVCLMFX-UHFFFAOYSA-N
70311
CCC(=O)C1=C(C=CC(=C1)C)O
C10H12O2
MFCD01098830
2-Propionyl-p-cresol
1-(2-hydroxy-5-methylphenyl)propan-1-one

Specifications

Yellow
1g
C10H12O2
2-Propionyl-p-cresol
CCC(=O)C1=C(C=CC(=C1)C)O
164.204
164.20
≥98.0% (GC)
2°C
938-45-4
MFCD01098830
CXZJBPYDVCLMFX-UHFFFAOYSA-N
1-(2-hydroxy-5-methylphenyl)propan-1-one
70311
Clear Liquid at 20°C
2′-Hydroxy-5′-methylpropiophenone
Safety and Handling

Safety and Handling

TSCA : Yes