Please login to your online account to display your discounted pricing

2-Hydroxy-5-methylbenzophenone, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-5-methylbenzophenone
CAS 1470-57-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H12O2
Linear Formula HOC6H3(CH3)COC6H5
Formula Weight 212.25
MDL Number MFCD00002379
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121430010

  SDS
thermo fisher scientific
AC121430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 212.25
Physical Form Crystalline Powder
Quantity 1g
Color Yellow
TSCA TSCA
EINECS Number 216-002-1
Melting Point 83° to 85°C
Beilstein 08, 177
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C14H12O2
MDL Number MFCD00002379
CAS 1470-57-1
Linear Formula HOC6H3(CH3)COC6H5
Chemical Name or Material 2-Hydroxy-5-methylbenzophenone
Assay Percent Range 99%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.