missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Hydroxy-5'-methylacetophenone 98.0+%, TCI America™
Supplier: TCI America H07905G
Specifications
2′-Hydroxy-5′-methylacetophenone | |
47°C | |
C9H10O2 | |
5 g | |
YNPDFBFVMJNGKZ-UHFFFAOYSA-N | |
1-(2-hydroxy-5-methylphenyl)ethanone | |
15068 | |
≥98.0% (GC) |
1450-72-2 | |
Yellow | |
MFCD00002380 | |
2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen | |
CC1=CC(=C(C=C1)O)C(=O)C | |
150.177 | |
150.18 | |
Crystalline Powder |
Chemical Identifiers
1450-72-2 | |
150.177 | |
YNPDFBFVMJNGKZ-UHFFFAOYSA-N | |
15068 | |
CC1=CC(=C(C=C1)O)C(=O)C |
C9H10O2 | |
MFCD00002380 | |
2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen | |
1-(2-hydroxy-5-methylphenyl)ethanone |
Safety and Handling
EINECSNumber : (3)-2937
TSCA : Yes