Please login to your online account to display your discounted pricing

2'-Hydroxy-5'-methylacetophenone 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  121380500

 View more versions of this product

Catalog No. AC121380500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 150.18
Assay ≥97.5%
Quantity 50g
Color Yellow
Beilstein 08, 111
TSCA TSCA
EINECS Number 215-915-2
Melting Point 45° to 49°C
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
Assay Percent Range 98%
MDL Number MFCD00002380
Molecular Formula C9 H10 O2
Name Note 98%
CAS 1450-72-2

Specifications

Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
Name Note 98%
CAS 1450-72-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H10 O2
Formula Weight 150.18
MDL Number MFCD00002380
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents