Please login to your online account to display your discounted pricing

2'-Hydroxy-5'-methylacetophenone 98%, ACROS Organics™

$341.69 - $341.69

Chemical Identifiers

Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
Name Note 98%
CAS 1450-72-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H10 O2
Formula Weight 150.18
MDL Number MFCD00002380
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121380500

  SDS
thermo fisher scientific
121380500
Glass bottle 50g Each for $341.69
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 150.18
Quantity 50g
Color Yellow
Beilstein 08, 111
EINECS Number 215-915-2
TSCA TSCA
Melting Point 45° to 49°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9 H10 O2
MDL Number MFCD00002380
CAS 1450-72-2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.