missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Hydroxy-5'-methylacetophenone, 98%

$72.04 - $677.28

Chemical Identifiers

CAS 1450-72-2
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00002380
Beilstein 1364840
Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1313606 SDS Alfa Aesar™
A1313606
5g Each for $72.04
AAA1313614 SDS Alfa Aesar™
A1313614
25g Each for $203.19
AAA1313622 SDS Alfa Aesar™
A1313622
100g Each for $677.28
Specifications

Description

Applications
2′-Hydroxy-5′-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

1450-72-2
150.18
1364840
C9H10O2
MFCD00002380
2'-Hydroxy-5'-methylacetophenone

Specifications

>110°C (230°F)
45° to 48°C
1450-72-2
150.18
1364840
98%
112° to 114°C (12mmHg)
5g
C9H10O2
MFCD00002380
2'-Hydroxy-5'-methylacetophenone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA