missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2′-Hydroxy-5′-methyl-3′-nitroacetophenone, TCI America™

$50.42 - $177.22

Chemical Identifiers

CAS 66108-30-3
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00192216
InChI Key XSHQMMIEZHWNAK-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methyl-3'-nitroacetophenone, 1-2-hydroxy-5-methyl-3-nitrophenyl ethanone, 2-hydroxy-5-methyl-3-nitroacetophenone, 2-acetyl-4-methyl-6-nitrophenol, ethanone,1-2-hydroxy-5-methyl-3-nitrophenyl, 1-acetyl-2-hydroxy-5-methyl-3-nitrobenzene, 2'-hydroxy-3'-nitro-5'-methylacetophenone, 1-2-hydroxy-5-methyl-3-nitro-phenyl-ethanone, 1-5-methyl-3-nitro-2-oxidanyl-phenyl ethanone, 1-2-hydroxy-5-methyl-3-nitrophenyl-1-ethanone
PubChem CID 736162
SMILES CC1=CC(=C(C(=C1)C(=O)C)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14795G View Documents TCI America
H14795G
5g Each for $50.42
 
H147925G View Documents TCI America
H147925G
25g Each for $177.22
 
Specifications

Chemical Identifiers

66108-30-3
195.174
XSHQMMIEZHWNAK-UHFFFAOYSA-N
736162
C9H9NO4
MFCD00192216
2'-hydroxy-5'-methyl-3'-nitroacetophenone, 1-2-hydroxy-5-methyl-3-nitrophenyl ethanone, 2-hydroxy-5-methyl-3-nitroacetophenone, 2-acetyl-4-methyl-6-nitrophenol, ethanone,1-2-hydroxy-5-methyl-3-nitrophenyl, 1-acetyl-2-hydroxy-5-methyl-3-nitrobenzene, 2'-hydroxy-3'-nitro-5'-methylacetophenone, 1-2-hydroxy-5-methyl-3-nitro-phenyl-ethanone, 1-5-methyl-3-nitro-2-oxidanyl-phenyl ethanone, 1-2-hydroxy-5-methyl-3-nitrophenyl-1-ethanone
CC1=CC(=C(C(=C1)C(=O)C)O)[N+](=O)[O-]

Specifications

Yellow
5g
C9H9NO4
2'-hydroxy-5'-methyl-3'-nitroacetophenone, 1-2-hydroxy-5-methyl-3-nitrophenyl ethanone, 2-hydroxy-5-methyl-3-nitroacetophenone, 2-acetyl-4-methyl-6-nitrophenol, ethanone,1-2-hydroxy-5-methyl-3-nitrophenyl, 1-acetyl-2-hydroxy-5-methyl-3-nitrobenzene, 2'-hydroxy-3'-nitro-5'-methylacetophenone, 1-2-hydroxy-5-methyl-3-nitro-phenyl-ethanone, 1-5-methyl-3-nitro-2-oxidanyl-phenyl ethanone, 1-2-hydroxy-5-methyl-3-nitrophenyl-1-ethanone
CC1=CC(=C(C(=C1)C(=O)C)O)[N+](=O)[O-]
736162
Crystal-Powder at 20°C
2′-Hydroxy-5′-methyl-3′-nitroacetophenone
134°C
66108-30-3
MFCD00192216
XSHQMMIEZHWNAK-UHFFFAOYSA-N
195.174
195.17
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No