Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Hydroxy-5'-methyl-3'-nitroacetophenone, 99%

$52.49 - $164.70

Chemical Identifiers

Chemical Name or Material 2'-Hydroxy-5'-methyl-3'-nitroacetophenone
CAS 66108-30-3
Molecular Formula C9H9NO4
Formula Weight 195.18
MDL Number MFCD00192216
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2767106

  SDS
alfa aesar™
H2767106
5g Each for $52.49

AAH2767114

  SDS
alfa aesar™
H2767114
25g Each for $164.70
Description & Specifications

Specifications

Formula Weight 195.18
Chemical Name or Material 2'-Hydroxy-5'-methyl-3'-nitroacetophenone
Assay 99%
Molecular Formula C9H9NO4
MDL Number MFCD00192216
CAS 66108-30-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 133° to 136°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.