Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-5-methoxybenzoic acid, 99%

$47.46 - $149.03

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-5-methoxybenzoic acid
CAS 2612-02-4
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD00002459
Beilstein 2209647
Synonym 5-Methoxysalicylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1627006

  SDS
alfa aesar™
L1627006
5g Each for $47.46

AAL1627014

  SDS
alfa aesar™
L1627014
25g Each for $149.03
Description & Specifications

Specifications

Formula Weight 168.15
Chemical Name or Material 2-Hydroxy-5-methoxybenzoic acid
MDL Number MFCD00002459
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 5-Methoxysalicylic acid
Health Hazard 1 Warning
CAS 2612-02-4
TSCA TSCA
Molecular Formula C8H8O4
Assay 99%
Melting Point 141° to 144°C
Beilstein 2209647
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.