missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-5-methoxybenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC164160500

 View more versions of this product

Catalog No. AC164160500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Hydroxy-5-methoxybenzaldehyde
Authentic
1.5770 to 1.5790 (20°C, 589nm)
50g
97.5
98%
HOC6H3(OCH3)CHO
08, 244
FZHSPPYCNDYIKD-UHFFFAOYSA-N
2-hydroxy-5-methoxybenzaldehyde
95695
Liquid
1.21g/mL
Glass bottle
Yellow
672-13-9
100.0
C8H8O3
MFCD00003332
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
COC1=CC(=C(C=C1)O)C=O
152.149
152.15
≥97.5%

Chemical Identifiers

672-13-9
152.149
FZHSPPYCNDYIKD-UHFFFAOYSA-N
95695
COC1=CC(=C(C=C1)O)C=O
C8H8O3
MFCD00003332
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
2-hydroxy-5-methoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents