Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-5-methoxybenzaldehyde, 98%

$96.74 - $858.85

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
CAS 672-13-9
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00003332
Beilstein 1100532
Synonym 5-Methoxysalicylaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1575306

  SDS
alfa aesar™
A1575306
5g Each for $96.74

AAA1575314

  SDS
alfa aesar™
A1575314
25g Each for $256.15

AAA1575318

  SDS
alfa aesar™
A1575318
50g Each for $465.59

AAA1575322

  SDS
alfa aesar™
A1575322
100g Each for $858.85
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Assay 98%
Molecular Formula C8H8O3
MDL Number MFCD00003332
CAS 672-13-9
Beilstein 1100532
Synonym 5-Methoxysalicylaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BZ2800000
Flash Point >110°C (230°F)
Boiling Point 248° to 252°C
Melting Point 13° to 16°C
Quantity 5g

Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.