missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Hydroxy-5'-methoxyacetophenone, 99%

$70.64 - $627.86

Chemical Identifiers

CAS 705-15-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008731
InChI Key MLIBGOFSXXWRIY-UHFFFAOYSA-N
Synonym 1-2-hydroxy-5-methoxy-phenyl-ethanone, 1-2-hydroxy-5-methoxyphenyl ethan-1-one, 1-2-hydroxy-5-methoxyphenyl ethanone, 1-acetyl-2-hydroxy-5-methoxybenzene, 2'-hydroxy-5'-methoxyacetophenone, 2'-hydroxy-5'-methoxyacetylphenone, 2-hydroxy-5-methoxyacetophenone, 5-methoxy-2-hydroxyacetophenone, acetophenone, 2'-hydroxy-5'-methoxy, ethanone, 1-2-hydroxy-5-methoxyphenyl
PubChem CID 69714
IUPAC Name 1-(2-hydroxy-5-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)OC)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1921606
View Documents
Alfa Aesar™
A1921606
5g
Each for $70.64
 
AAA1921614
View Documents
Alfa Aesar™
A1921614
25g
Each for $229.42
 
AAA1921622
View Documents
Alfa Aesar™
A1921622
100g
Each for $627.86
 
Description

Description

Specifications

Chemical Identifiers

705-15-7
166.176
MLIBGOFSXXWRIY-UHFFFAOYSA-N
69714
CC(=O)C1=C(C=CC(=C1)OC)O
C9H10O3
MFCD00008731
1-2-hydroxy-5-methoxy-phenyl-ethanone, 1-2-hydroxy-5-methoxyphenyl ethan-1-one, 1-2-hydroxy-5-methoxyphenyl ethanone, 1-acetyl-2-hydroxy-5-methoxybenzene, 2'-hydroxy-5'-methoxyacetophenone, 2'-hydroxy-5'-methoxyacetylphenone, 2-hydroxy-5-methoxyacetophenone, 5-methoxy-2-hydroxyacetophenone, acetophenone, 2'-hydroxy-5'-methoxy, ethanone, 1-2-hydroxy-5-methoxyphenyl
1-(2-hydroxy-5-methoxyphenyl)ethanone

Specifications

>110°C (230°F)
5g
C9H10O3
1239364
MLIBGOFSXXWRIY-UHFFFAOYSA-N
1-(2-hydroxy-5-methoxyphenyl)ethanone
69714
99%
48°C to 50°C
705-15-7
MFCD00008731
1-2-hydroxy-5-methoxy-phenyl-ethanone, 1-2-hydroxy-5-methoxyphenyl ethan-1-one, 1-2-hydroxy-5-methoxyphenyl ethanone, 1-acetyl-2-hydroxy-5-methoxybenzene, 2'-hydroxy-5'-methoxyacetophenone, 2'-hydroxy-5'-methoxyacetylphenone, 2-hydroxy-5-methoxyacetophenone, 5-methoxy-2-hydroxyacetophenone, acetophenone, 2'-hydroxy-5'-methoxy, ethanone, 1-2-hydroxy-5-methoxyphenyl
CC(=O)C1=C(C=CC(=C1)OC)O
166.176
166.18
2'-Hydroxy-5'-methoxyacetophenone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-882-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications