Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-5-iodo-3-methoxybenzaldehyde, 97%

$86.19 - $319.43

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-5-iodo-3-methoxybenzaldehyde
CAS 7359-14-0
Molecular Formula C7H8IO3
Formula Weight 278.05
MDL Number MFCD00016594
Synonym 5-Iodo-o-vanillin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2481103

  SDS
alfa aesar™
B2481103
1g Each for $86.19

AAB2481106

  SDS
alfa aesar™
B2481106
5g Each for $319.43

AAB2481114

  SDS
alfa aesar™
B2481114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Hydroxy-5-iodo-3-methoxybenzaldehyde
Melting Point 127° to 130°C
Quantity 1g
CAS 7359-14-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C7H8IO3
Formula Weight 278.05
MDL Number MFCD00016594
Synonym 5-Iodo-o-vanillin
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.