missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-(trifluoromethyl)pyrimidine, 99%

$44.41 - $197.39

Chemical Identifiers

CAS 104048-92-2
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00192529
InChI Key WCEBFEVZTGLOHC-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am
PubChem CID 1240050
IUPAC Name 6-(trifluoromethyl)-1H-pyrimidin-2-one
SMILES C1=C(NC(=O)N=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6015103 View Documents Alfa Aesar™
H6015103
1g Each for $44.41
 
AAH6015106 View Documents Alfa Aesar™
H6015106
5g Each for $197.39
 
Specifications

Chemical Identifiers

104048-92-2
164.087
WCEBFEVZTGLOHC-UHFFFAOYSA-N
1240050
C1=C(NC(=O)N=C1)C(F)(F)F
C5H3F3N2O
MFCD00192529
2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am
6-(trifluoromethyl)-1H-pyrimidin-2-one

Specifications

223°C to 227°C
104048-92-2
MFCD00192529
WCEBFEVZTGLOHC-UHFFFAOYSA-N
6-(trifluoromethyl)-1H-pyrimidin-2-one
1240050
99%
1g
C5H3F3N2O
2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am
C1=C(NC(=O)N=C1)C(F)(F)F
164.087
164.09
2-Hydroxy-4-(trifluoromethyl)pyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning