Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-(trifluoromethyl)pyridine, 98%

$236.57 - $658.46

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyridine
CAS 50650-59-4
Molecular Formula C6H4F3NO
Formula Weight 163.1
MDL Number MFCD00153195
Synonym 4-Trifluoromethyl-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2575603

  SDS
alfa aesar™
H2575603
1g Each for $658.46

AAH25756MD

  SDS
alfa aesar™
H25756MD
250mg Each for $236.57
Description & Specifications

Specifications

Formula Weight 163.1
Health Hazard 1 Warning
Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 160°C
Molecular Formula C6H4F3NO
Synonym 4-Trifluoromethyl-2(1H)-pyridone
MDL Number MFCD00153195
Quantity 1g
CAS 50650-59-4
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
2-Hydroxy-4-(trifluoromethyl)pyridine is used in the production of agrochemicals and pharmaceuticals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.