missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-n-octyloxybenzophenone 98.0+%, TCI America™
$42.87 - $260.97
Chemical Identifiers
| CAS | 1843-05-6 |
|---|---|
| Molecular Formula | C21H26O3 |
| Molecular Weight (g/mol) | 326.436 |
| MDL Number | MFCD00027327 |
| InChI Key | QUAMTGJKVDWJEQ-UHFFFAOYSA-N |
| Synonym | octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130 |
| PubChem CID | 15797 |
| IUPAC Name | (2-hydroxy-4-octoxyphenyl)-phenylmethanone |
| SMILES | CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
Chemical Identifiers
| 1843-05-6 | |
| 326.436 | |
| QUAMTGJKVDWJEQ-UHFFFAOYSA-N | |
| 15797 | |
| CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
| C21H26O3 | |
| MFCD00027327 | |
| octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130 | |
| (2-hydroxy-4-octoxyphenyl)-phenylmethanone |
Specifications
| 1843-05-6 | |
| White-Yellow | |
| MFCD00027327 | |
| octabenzone, 2-hydroxy-4-n-octyloxybenzophenone, benzophenone-12, 2-hydroxy-4-octyloxy phenyl phenyl methanone, benzon oo, 2-hydroxy-4-octyloxy benzophenone, benzophenone 12, cyasorb uv 531, spectra-sorb uv 531, sumisorb 130 | |
| CCCCCCCCOC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O | |
| 326.436 | |
| 326.44 | |
| Crystalline Powder |
| 48°C | |
| C21H26O3 | |
| 25 g | |
| QUAMTGJKVDWJEQ-UHFFFAOYSA-N | |
| (2-hydroxy-4-octoxyphenyl)-phenylmethanone | |
| 15797 | |
| ≥98.0% (GC) | |
| 2-Hydroxy-4-n-octyloxybenzophenone |
Safety and Handling
EINECSNumber : (4)-0141
RTECSNumber : DJ1595000
TSCA : Yes