Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-n-octyloxybenzophenone, 99%

$27.85 - $56.04

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4-n-octyloxybenzophenone
CAS 1843-05-6
Molecular Formula C21H26O3
Formula Weight 326.44
MDL Number MFCD00027327
Beilstein 1915198
Merck Index 14,6742
Synonym Octabenzone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1175014

 
alfa aesar™
L1175014
25g Each for $27.85

AAL1175022

 
alfa aesar™
L1175022
100g Each for $56.04
Description & Specifications

Specifications

Formula Weight 326.44
Chemical Name or Material 2-Hydroxy-4-n-octyloxybenzophenone
TSCA TSCA
Molecular Formula C21H26O3
MDL Number MFCD00027327
CAS 1843-05-6
Synonym Octabenzone
Melting Point 46° to 49°C
Assay 99%
Beilstein 1915198
Merck Index 14,6742
RTECS Number DJ1595000
Quantity 25g