Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-methylquinoline, 98%

$80.24 - $80.24

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4-methylquinoline
CAS 607-66-9
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD00006745
Beilstein 118331
Synonym 2-Hydroxylepidine; 4-Methylcarbostyril
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0741714

 
alfa aesar™
L0741714
25g Each for $80.24
Description & Specifications

Specifications

Chemical Name or Material 2-Hydroxy-4-methylquinoline
Melting Point 223° to 228°C
Quantity 25g
CAS 607-66-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD00006745
Beilstein 118331
Synonym 2-Hydroxylepidine; 4-Methylcarbostyril