Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-methylpyrimidine hydrochloride, 97%

$333.00 - $333.00

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4-methylpyrimidine hydrochloride
CAS 5348-51-6
Molecular Formula C5H6N2O·HCl
Formula Weight 146.58
MDL Number MFCD00151347
Beilstein 3691454
Synonym 4-Methylpyrimidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2225522

  SDS
alfa aesar™
B2225522
100g Each for $333.00
Description & Specifications

Specifications

Formula Weight 146.58
MDL Number MFCD00151347
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 2-Hydroxy-4-methylpyrimidine hydrochloride
Assay 97%
Molecular Formula C5H6N2O·HCl
CAS 5348-51-6
Odor Odorless
Quantity 100g
Synonym 4-Methylpyrimidinol hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈240°C (dec.)
Health Hazard 1 Warning
Beilstein 3691454
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.