Please login to your online account to display your discounted pricing

2-Hydroxy-4-methylbenzaldehyde, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC433140250

 View more versions of this product

Catalog No. AC433140250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 136.15
Assay ≥96.0%
Physical Form Crystalline Powder
Quantity 25g
Color White
Melting Point 59° to 63°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C8H8O2
Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
CAS 698-27-1
MDL Number MFCD00799550

Specifications

Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
CAS 698-27-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8O2
Formula Weight 136.15
MDL Number MFCD00799550
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents