missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methyl-5-nitropyridine, TCI America™

$85.56 - $253.61

Chemical Identifiers

CAS 21901-41-7
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00010690
InChI Key AIEHUZHKFUNHCJ-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
PubChem CID 345371
IUPAC Name 4-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)NC=C1[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10505G View Documents TCI America
H10505G
5g Each for $253.61
 
H10501G View Documents TCI America
H10501G
1g Each for $85.56
 
Specifications

Chemical Identifiers

21901-41-7
154.125
AIEHUZHKFUNHCJ-UHFFFAOYSA-N
345371
CC1=CC(=O)NC=C1[N+](=O)[O-]
C6H6N2O3
MFCD00010690
2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
4-methyl-5-nitro-1H-pyridin-2-one

Specifications

Undesignated
5g
C6H6N2O3
2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
CC1=CC(=O)NC=C1[N+](=O)[O-]
154.125
154.13
≥98.0% (GC,T)
190°C
21901-41-7
MFCD00010690
AIEHUZHKFUNHCJ-UHFFFAOYSA-N
4-methyl-5-nitro-1H-pyridin-2-one
345371
Crystal-Powder at 20°C
2-Hydroxy-4-methyl-5-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No