missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methyl-3-nitropyridine, TCI America™

$62.64 - $192.50

Chemical Identifiers

CAS 21901-18-8
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
InChI Key HZCWTTHQRMHIIE-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
PubChem CID 345370
IUPAC Name 4-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=C(C(=O)NC=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H11705G View Documents TCI America
H11705G
5g Each for $192.50
 
H11701G View Documents TCI America
H11701G
1g Each for $62.64
 
Specifications

Chemical Identifiers

21901-18-8
154.125
2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
4-methyl-3-nitro-1H-pyridin-2-one
C6H6N2O3
HZCWTTHQRMHIIE-UHFFFAOYSA-N
345370
CC1=C(C(=O)NC=C1)[N+](=O)[O-]

Specifications

Yellow
5g
C6H6N2O3
HZCWTTHQRMHIIE-UHFFFAOYSA-N
4-methyl-3-nitro-1H-pyridin-2-one
345370
Crystal-Powder at 20°C
2-Hydroxy-4-methyl-3-nitropyridine
234°C
21901-18-8
2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
CC1=C(C(=O)NC=C1)[N+](=O)[O-]
154.125
154.13
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No