missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methoxybenzophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121360050

 View more versions of this product

Catalog No. AC121360050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Hydroxy-4-methoxybenzophenone
131-57-7
100.0
White to Yellow
>100°C
97.5% min. (GC)
C14H12O3
MFCD00008387
08,312
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
DXGLGDHPHMLXJC-UHFFFAOYSA-N
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
4632
228.25
Crystalline Powder
98%
97.5
>62.5°C
150°C to 160°C (5.0 mmHg)
Authentic
Glass bottle
HOC6H3(OCH3)COC6H5
5 g
15,7053
Solubility in water: .. Other solubilities: readily soluble in most organic solvents
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
228.25
CHEBI:34283
98%

Chemical Identifiers

131-57-7
228.25
DXGLGDHPHMLXJC-UHFFFAOYSA-N
4632
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
C14H12O3
MFCD00008387
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
CHEBI:34283
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
Safety and Handling

Safety and Handling

EINECSNumber : 205-031-5

RTECSNumber : DJ1575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only