missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methoxybenzophenone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC121360010

 View more versions of this product

Catalog No. AC121360010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Hydroxy-4-methoxybenzophenone
Authentic
150°C to 160°C (5.0mmHg)
>62.5°C
98%
97.5
97.5% min. (GC)
HOC6H3(OCH3)COC6H5
08,312
2-benzoyl-5-methoxyphenol, 2-hydroxy-4-methoxybenzophenone, 2-hydroxy-4-methoxyphenyl phenyl methanone, 4-methoxy-2-hydroxybenzophenone, anuvex, benzophenone-3, oxibenzona, oxybenzon, oxybenzone, oxybenzonum
DXGLGDHPHMLXJC-UHFFFAOYSA-N
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
4632
228.25
98%
>100°C
Glass bottle
White to Yellow
1kg
131-57-7
100.0
C14H12O3
MFCD00008387
15,7053
Solubility in water: .. Other solubilities: readily soluble in most organic solvents
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
228.25
CHEBI:34283
Crystalline Powder

Chemical Identifiers

131-57-7
228.25
DXGLGDHPHMLXJC-UHFFFAOYSA-N
4632
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
C14H12O3
MFCD00008387
2-benzoyl-5-methoxyphenol, 2-hydroxy-4-methoxybenzophenone, 2-hydroxy-4-methoxyphenyl phenyl methanone, 4-methoxy-2-hydroxybenzophenone, anuvex, benzophenone-3, oxibenzona, oxybenzon, oxybenzone, oxybenzonum
CHEBI:34283
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
Safety and Handling

Safety and Handling

EINECSNumber : 205-031-5

RTECSNumber : DJ1575000

TSCA : TSCA

SDS Documents