Please login to your online account to display your discounted pricing

2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 98%, ACROS Organics™


Quantity 50g

Manufacturer: thermo fisher scientific  AC358060500

 View more versions of this product

Catalog No. AC358060500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 308.31
Physical Form Powder
Quantity 50g
Color White to yellow
Merck Index 15, 9113
CAS Max % 100.0
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Water 3% max. (K.F.)
Infrared Spectrum Authentic
Name Note 98%
CAS 4065-45-6
Molecular Formula C14H12O6S
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid
Name Note 98%
CAS 4065-45-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H12O6S
Formula Weight 308.31
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents