missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 98%, ACROS Organics™

$263.10 - $263.10

Chemical Identifiers

CAS 4065-45-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C14H12O6S
Formula Weight 308.31
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC358060500 SDS Thermo Fisher Scientific
AC358060500
50g Each for $263.10
Specifications Description & Specifications

Chemical Identifiers

4065-45-6
100.0
C14H12O6S
2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 98%
97.5
98%
308.31
≥97.5%

Specifications

White to yellow
4065-45-6
100.0
Powder
C14H12O6S
3% max. (K.F.)
2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 98%
50g
97.5
Authentic
98%
308.31
15, 9113
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.