missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4-methoxybenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H5534503

 View more versions of this product

Catalog No. AAH5534503



Specifications

Specifications

2-Hydroxy-4-methoxybenzoic acid
1g
C8H8O4
2-hydroxy-4-methoxy-benzoic acid
COC1=CC(=C(C=C1)C(=O)O)O
168.148
168.15
158°C to 159°C
2237-36-7
MFCD00002450
MRIXVKKOHPQOFK-UHFFFAOYSA-N
2-hydroxy-4-methoxybenzoic acid
75231
98%

Chemical Identifiers

2237-36-7
168.148
MRIXVKKOHPQOFK-UHFFFAOYSA-N
75231
COC1=CC(=C(C=C1)C(=O)O)O
C8H8O4
MFCD00002450
2-hydroxy-4-methoxy-benzoic acid
2-hydroxy-4-methoxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 218-801-0

RTECSNumber : DH2270000

TSCA : No

Recommended Storage : Ambient temperatures

Documents