missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-methoxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America H069925G
Specifications
| 2-Hydroxy-4-methoxybenzaldehyde | |
| 42°C | |
| MFCD00003327 | |
| 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw | |
| COC1=CC=C(C=O)C(O)=C1 | |
| 152.15 | |
| 152.15 | |
| Crystalline Powder |
| 673-22-3 | |
| C8H8O3 | |
| 25 g | |
| WZUODJNEIXSNEU-UHFFFAOYSA-N | |
| 2-hydroxy-4-methoxybenzaldehyde | |
| 69600 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 673-22-3 | |
| 152.15 | |
| WZUODJNEIXSNEU-UHFFFAOYSA-N | |
| 69600 | |
| COC1=CC=C(C=O)C(O)=C1 |
| C8H8O3 | |
| MFCD00003327 | |
| 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw | |
| 2-hydroxy-4-methoxybenzaldehyde |
Safety and Handling
RTECSNumber : BZ2810000
TSCA : Yes