Learn More
2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2244522
Description
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.
Solubility
Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
2-Hydroxy-4-(methacryloyloxy)benzophenone | |
77°C to 79°C | |
C17H14O4 | |
100 g | |
4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenone | |
IMNBHNRXUAJVQE-UHFFFAOYSA-N | |
(4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate | |
74856 | |
99% |
2035-72-5 | |
1.18 | |
MFCD00080561 | |
Light sensitive | |
Insoluble in water. | |
CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O | |
282.295 | |
282.3 |
Chemical Identifiers
2035-72-5 | |
282.295 | |
IMNBHNRXUAJVQE-UHFFFAOYSA-N | |
74856 | |
CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
C17H14O4 | |
MFCD00080561 | |
4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenone | |
(4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-000-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only