Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4,6-dimethylpyrimidine, 97%

$91.46 - $207.93

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4,6-dimethylpyrimidine
CAS 108-79-2
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD00006072
Synonym 4,6-Dimethyl-2-hydroxypyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1367614

  SDS
alfa aesar™
A1367614
25g Each for $91.46

AAA1367622

  SDS
alfa aesar™
A1367622
100g Each for $207.93
Description & Specifications

Specifications

Formula Weight 124.14
Assay 97%
Quantity 25g
TSCA TSCA
Synonym 4,6-Dimethyl-2-hydroxypyrimidine
Molecular Formula C6H8N2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 108-79-2
Health Hazard 1 Warning
Chemical Name or Material 2-Hydroxy-4,6-dimethylpyrimidine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00006072
Melting Point 202° to 204°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.