Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-4,6-dimethoxybenzoic acid, 97%

$110.69 - $380.20

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-4,6-dimethoxybenzoic acid
CAS 3187-19-7
Molecular Formula C9H10O5
Formula Weight 198.17
MDL Number MFCD00013977
Beilstein 2213838
Synonym 4,6-Dimethoxy-2-hydroxybenzoic acid; 4,6-Dimethoxysalicylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0138803

  SDS
alfa aesar™
L0138803
1g Each for $110.69

AAL0138806

  SDS
alfa aesar™
L0138806
5g Each for $380.20
Description & Specifications

Specifications

Formula Weight 198.17
Chemical Name or Material 2-Hydroxy-4,6-dimethoxybenzoic acid
Synonym 4,6-Dimethoxy-2-hydroxybenzoic acid; 4,6-Dimethoxysalicylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD00013977
CAS 3187-19-7
Beilstein 2213838
Health Hazard 1 Warning
Quantity 1g
Molecular Formula C9H10O5
Melting Point 160° to 162°C

Applications
2-Hydroxy-4,6-dimethoxybenzoic acid is a reagent used in the synthesis of rac Zearalenone-d6. rac Zearalenone-d6 s an estrogenic mycotoxin produced by Fusarium fungi commonly found in grains. One of a group of compounds known as resorcylic acid lactones.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.