missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%

Manufacturer:  Alfa Aesar™ A1458506

 View more versions of this product

Catalog No. AAA1458506



Description

Description

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2'-Hydroxy-4',6'-dimethoxyacetophenone
80°C to 83°C
90-24-4
MFCD00017243
14,10067
FBUBVLUPUDBFME-UHFFFAOYSA-N
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
66654
196.2
185°C (20mmHg)
5g
C10H12O4
1285013
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
CC(=O)C1=C(C=C(C=C1OC)OC)O
196.202
CHEBI:10070
98%

Chemical Identifiers

90-24-4
196.202
FBUBVLUPUDBFME-UHFFFAOYSA-N
66654
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
C10H12O4
MFCD00017243
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
CHEBI:10070
CC(=O)C1=C(C=C(C=C1OC)OC)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents