missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ), TCI America™
$50.42 - $192.50
Chemical Identifiers
CAS | 16969-10-1 |
---|---|
Molecular Formula | C12H14O4 |
Molecular Weight (g/mol) | 222.24 |
MDL Number | MFCD00066300 |
InChI Key | HHQAGBQXOWLTLL-UHFFFAOYSA-N |
Synonym | Acrylic Acid 2-Hydroxy-3-phenoxypropyl Ester |
PubChem CID | 4443516 |
IUPAC Name | (2-hydroxy-3-phenoxypropyl) prop-2-enoate |
SMILES | C=CC(=O)OCC(COC1=CC=CC=C1)O |
Chemical Identifiers
16969-10-1 | |
222.24 | |
HHQAGBQXOWLTLL-UHFFFAOYSA-N | |
4443516 | |
C=CC(=O)OCC(COC1=CC=CC=C1)O |
C12H14O4 | |
MFCD00066300 | |
Acrylic Acid 2-Hydroxy-3-phenoxypropyl Ester | |
(2-hydroxy-3-phenoxypropyl) prop-2-enoate |
Specifications
Yellow | |
16969-10-1 | |
MFCD00066300 | |
HHQAGBQXOWLTLL-UHFFFAOYSA-N | |
(2-hydroxy-3-phenoxypropyl) prop-2-enoate | |
4443516 | |
Clear Liquid at 20°C | |
2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ) |
25g | |
C12H14O4 | |
Acrylic Acid 2-Hydroxy-3-phenoxypropyl Ester | |
C=CC(=O)OCC(COC1=CC=CC=C1)O | |
222.24 | |
222.24 | |
≥85.0% (GC) |
Safety and Handling
EINECSNumber : (3)-0565
TSCA : Yes