Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-nitropyridine, 98%

$73.38 - $73.38

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3-nitropyridine
CAS 6332-56-5
Molecular Formula C5H4N2O3
Formula Weight 140.1
MDL Number MFCD00006270
Beilstein 124474
Synonym 3-Nitro-2-pyridinol; 3-Nitro-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1776806

  SDS
alfa aesar™
A1776806
5g Each for $73.38
Description & Specifications

Specifications

Formula Weight 140.1
Chemical Name or Material 2-Hydroxy-3-nitropyridine
Assay 98%
Molecular Formula C5H4N2O3
MDL Number MFCD00006270
CAS 6332-56-5
Beilstein 124474
Synonym 3-Nitro-2-pyridinol; 3-Nitro-2(1H)-pyridone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UU7718000
Melting Point ≈220°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.