missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine, 97%

Manufacturer:  Alfa Aesar™ H6369406

 View more versions of this product

Catalog No. AAH6369406



Specifications

Specifications

2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine
189°C to 190°C
33252-64-1
MFCD00153193
2-hydroxy-3-nitro-5-trifluoromethyl pyridine, 3-nitro-5-trifluoromethyl pyridin-2 1h-one, 3-nitro-5-trifluoromethyl pyridin-2-ol, 3-nitro-5-trifluoromethyl-2-pyridinone, 2 1h-pyridinone,3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitropyridin-2-ol, 3-nitro-5-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 3-nitro-5-trifluoromethyl, 3-nitro-5-trifluoromethyl hydropyridin-2-one, pubchem9284
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
208.096
208.09
97%
White
5g
C6H3F3N2O3
UN2811
JYXKHKBZLLIWEV-UHFFFAOYSA-N
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
2775093
Solid

Chemical Identifiers

33252-64-1
208.096
JYXKHKBZLLIWEV-UHFFFAOYSA-N
2775093
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
C6H3F3N2O3
MFCD00153193
2-hydroxy-3-nitro-5-trifluoromethyl pyridine, 3-nitro-5-trifluoromethyl pyridin-2 1h-one, 3-nitro-5-trifluoromethyl pyridin-2-ol, 3-nitro-5-trifluoromethyl-2-pyridinone, 2 1h-pyridinone,3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitropyridin-2-ol, 3-nitro-5-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 3-nitro-5-trifluoromethyl, 3-nitro-5-trifluoromethyl hydropyridin-2-one, pubchem9284
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents