Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate

Chemical Identifiers

Chemical Name or Material (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate
MDL Number MFCD00078139
Synonym Quaternary ammonium beta-cylodextrin; QA-beta-CD
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2760403

 
alfa aesar™
H2760403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27604MD

 
alfa aesar™
H27604MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate
MDL Number MFCD00078139
Synonym Quaternary ammonium beta-cylodextrin; QA-beta-CD
Melting Point ≈280°C
Quantity 1g
RTECS Number GU2293000