missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-3-methylpyrazine 98.0+%, TCI America™

$210.67 - $709.38

Chemical Identifiers

CAS 19838-07-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD01646120
InChI Key LDQRWMQHTORUIY-UHFFFAOYSA-N
Synonym 2 1h-pyrazinone, 3-methyl, 2 1h-pyrazinone,3-methyl, 2-hydroxy-3-methylpyrazine, 3-methyl-2 1h-pyrazinone, 3-methyl-2-hydroxypyrazine, 3-methyl-2-pyrazinol, 3-methylpyrazin-2 1h-one, 3-methylpyrazin-2-ol, unii-s951qmj75e
PubChem CID 88276
IUPAC Name 3-methyl-1H-pyrazin-2-one
SMILES CC1=NC=CNC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15191G View Documents Tci America
H15191G
1g Each for $210.67
 
H15195G View Documents Tci America
H15195G
5g Each for $709.38
 
Specifications

Chemical Identifiers

19838-07-4
110.116
LDQRWMQHTORUIY-UHFFFAOYSA-N
88276
CC1=NC=CNC1=O
C5H6N2O
MFCD01646120
2 1h-pyrazinone, 3-methyl, 2 1h-pyrazinone,3-methyl, 2-hydroxy-3-methylpyrazine, 3-methyl-2 1h-pyrazinone, 3-methyl-2-hydroxypyrazine, 3-methyl-2-pyrazinol, 3-methylpyrazin-2 1h-one, 3-methylpyrazin-2-ol, unii-s951qmj75e
3-methyl-1H-pyrazin-2-one

Specifications

Yellow
1g
C5H6N2O
2 1h-pyrazinone, 3-methyl, 2 1h-pyrazinone,3-methyl, 2-hydroxy-3-methylpyrazine, 3-methyl-2 1h-pyrazinone, 3-methyl-2-hydroxypyrazine, 3-methyl-2-pyrazinol, 3-methylpyrazin-2 1h-one, 3-methylpyrazin-2-ol, unii-s951qmj75e
CC1=NC=CNC1=O
110.116
110.12
≥98.0% (GC)
150°C
19838-07-4
MFCD01646120
LDQRWMQHTORUIY-UHFFFAOYSA-N
3-methyl-1H-pyrazin-2-one
88276
Crystal-Powder at 20°C
2-Hydroxy-3-methylpyrazine
Safety and Handling

Safety and Handling

TSCA : No