Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-methylpyrazine, 96%

$113.14 - $386.36

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3-methylpyrazine
CAS 19838-07-4
Molecular Formula C5H6N2O
Formula Weight 110.11
MDL Number MFCD01646120
Synonym 3-Methyl-2-pyrazinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5188003

  SDS
alfa aesar™
H5188003
1g Each for $386.36

AAH51880MD

  SDS
alfa aesar™
H51880MD
250mg Each for $113.14
Description & Specifications

Specifications

Formula Weight 110.11
Chemical Name or Material 2-Hydroxy-3-methylpyrazine
Assay 96%
Molecular Formula C5H6N2O
MDL Number MFCD01646120
CAS 19838-07-4
Synonym 3-Methyl-2-pyrazinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 152° to 153°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.