missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-methylbenzhydrazide, 98%

$76.50 - $270.92

Chemical Identifiers

CAS 30991-42-5
Molecular Formula C8H10N2O2
Formula Weight 166.18
MDL Number MFCD00017052
Beilstein 2639905
Synonym 3-Methylsalicylhydrazide
Chemical Name or Material 2-Hydroxy-3-methylbenzhydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0898003 SDS Alfa Aesar™
L0898003
1g Each for $76.50
AAL0898006 SDS Alfa Aesar™
L0898006
5g Each for $270.92
Specifications Description & Specifications

Chemical Identifiers

30991-42-5
166.18
2639905
2-Hydroxy-3-methylbenzhydrazide
C8H10N2O2
MFCD00017052
3-Methylsalicylhydrazide

Specifications

189° to 193°C
30991-42-5
166.18
2639905
2-Hydroxy-3-methylbenzhydrazide
1g
C8H10N2O2
MFCD00017052
3-Methylsalicylhydrazide
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning