missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-3-methyl-5-nitropyridine, TCI America™

$68.75 - $206.25

Chemical Identifiers

CAS 21901-34-8
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
InChI Key FPTYZBDNBMVYCL-UHFFFAOYSA-N
Synonym 2-hydroxy-3-methyl-5-nitropyridine, 3-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-3-picoline, 3-methyl-5-nitro-2-pyridone, 2-hydroxy-5-nitro-3-methylpyridine, 2 1h-pyridinone, 3-methyl-5-nitro, 3-methyl-5-nitro-2 1h-pyridinone, 3-methyl-5-nitro-2-pyridinol, 3-methyl-5-nitro-pyridin-2-ol, 6-hydroxy-5-methyl-3-nitropyridine
PubChem CID 89097
IUPAC Name 3-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=CC(=CNC1=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H117325G View Documents TCI America
H117325G
25g Each for $206.25
 
H11735G View Documents TCI America
H11735G
5g Each for $68.75
 
Specifications

Chemical Identifiers

21901-34-8
154.125
2-hydroxy-3-methyl-5-nitropyridine, 3-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-3-picoline, 3-methyl-5-nitro-2-pyridone, 2-hydroxy-5-nitro-3-methylpyridine, 2 1h-pyridinone, 3-methyl-5-nitro, 3-methyl-5-nitro-2 1h-pyridinone, 3-methyl-5-nitro-2-pyridinol, 3-methyl-5-nitro-pyridin-2-ol, 6-hydroxy-5-methyl-3-nitropyridine
3-methyl-5-nitro-1H-pyridin-2-one
C6H6N2O3
FPTYZBDNBMVYCL-UHFFFAOYSA-N
89097
CC1=CC(=CNC1=O)[N+](=O)[O-]

Specifications

Undesignated
25g
C6H6N2O3
FPTYZBDNBMVYCL-UHFFFAOYSA-N
3-methyl-5-nitro-1H-pyridin-2-one
89097
Crystal-Powder at 20°C
2-Hydroxy-3-methyl-5-nitropyridine
229°C
21901-34-8
2-hydroxy-3-methyl-5-nitropyridine, 3-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-3-picoline, 3-methyl-5-nitro-2-pyridone, 2-hydroxy-5-nitro-3-methylpyridine, 2 1h-pyridinone, 3-methyl-5-nitro, 3-methyl-5-nitro-2 1h-pyridinone, 3-methyl-5-nitro-2-pyridinol, 3-methyl-5-nitro-pyridin-2-ol, 6-hydroxy-5-methyl-3-nitropyridine
CC1=CC(=CNC1=O)[N+](=O)[O-]
154.125
154.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No