Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-methyl-5-nitropyridine, 97%

$57.41 - $203.42

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3-methyl-5-nitropyridine
CAS 21901-34-8
Molecular Formula C6H6N2O3
Formula Weight 154.13
MDL Number MFCD03095073
Beilstein 126949
Synonym 2-Hydroxy-5-nitro-3-picoline; 3-Methyl-5-nitro-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAL1986603

  SDS
alfa aesar™
L1986603
Each for $57.41

AAL1986606

  SDS
alfa aesar™
L1986606
Each for $203.42
Description & Specifications

Specifications

Formula Weight 154.13
Chemical Name or Material 2-Hydroxy-3-methyl-5-nitropyridine
MDL Number MFCD03095073
CAS 21901-34-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 97%
Molecular Formula C6H6N2O3
Beilstein 126949
Synonym 2-Hydroxy-5-nitro-3-picoline; 3-Methyl-5-nitro-2-pyridinol
Melting Point 228° to 231°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.