Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-methoxybenzaldehyde, 99%

$45.09 - $338.64

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3-methoxybenzaldehyde
CAS 148-53-8
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00003322
Beilstein 471913
Synonym 3-Methoxysalicylaldehyde; o-Vanillin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1567214

  SDS
alfa aesar™
A1567214
25g Each for $45.09

AAA1567222

  SDS
alfa aesar™
A1567222
100g Each for $126.08

AAA1567236

  SDS
alfa aesar™
A1567236
500g Each for $338.64
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material 2-Hydroxy-3-methoxybenzaldehyde
TSCA TSCA
Assay 99%
Molecular Formula C8H8O3
MDL Number MFCD00003322
CAS 148-53-8
Beilstein 471913
Synonym 3-Methoxysalicylaldehyde; o-Vanillin
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CU6530000
Boiling Point 265° to 266°C
Melting Point 40° to 42°C
Quantity 25g

Applications
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.