Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3-iodo-5-methylpyridine, 95%

$135.61 - $308.89

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3-iodo-5-methylpyridine
CAS 313678-93-2
Molecular Formula C6H6INO
Formula Weight 235.02
MDL Number MFCD09037469
Synonym 3-Iodo-5-methyl-2(1H)pyridone; 5-Iodo-3-picolin-6-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2775103

  SDS
alfa aesar™
H2775103
1g Each for $308.89

AAH27751MD

  SDS
alfa aesar™
H27751MD
250mg Each for $135.61
Description & Specifications

Specifications

Formula Weight 235.02
MDL Number MFCD09037469
Molecular Formula C6H6INO
Chemical Name or Material 2-Hydroxy-3-iodo-5-methylpyridine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
CAS 313678-93-2
Synonym 3-Iodo-5-methyl-2(1H)pyridone; 5-Iodo-3-picolin-6-ol
Melting Point 190° to 193°C

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.