Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-3,5-dinitropyridine, 98+%

$60.12 - $212.46

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-3,5-dinitropyridine
CAS 2980-33-8
Molecular Formula C5H3N3O5
Formula Weight 185.1
MDL Number MFCD00039716
Beilstein 196025
Synonym 3,5-Dinitro-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0791404

  SDS
alfa aesar™
L0791404
2g Each for $60.12

AAL0791409

  SDS
alfa aesar™
L0791409
10g Each for $212.46
Description & Specifications

Specifications

Formula Weight 185.1
TSCA TSCA
Chemical Name or Material 2-Hydroxy-3,5-dinitropyridine
Assay ≥98%
MDL Number MFCD00039716
Synonym 3,5-Dinitro-2-pyridinol
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2980-33-8
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 176° to 179°C
Beilstein 196025
Molecular Formula C5H3N3O5
Quantity 2g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.