missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-2-propanesulfonic acid monosodium salt, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03625636
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Hydroxy-2-propanesulfonic acid monosodium salt | |
C3H8NaO4S+ | |
500 g | |
YNJORDSKPXMABC-UHFFFAOYSA-N | |
sodium;2-hydroxypropane-2-sulfonic acid | |
54602011 | |
97% |
540-92-1 | |
MFCD00031395 | |
sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid | |
CC(C)(O)S(=O)(=O)O.[Na+] | |
163.143 | |
162.14 | |
Crystalline |
Chemical Identifiers
540-92-1 | |
163.143 | |
YNJORDSKPXMABC-UHFFFAOYSA-N | |
54602011 | |
CC(C)(O)S(=O)(=O)O.[Na+] |
C3H8NaO4S+ | |
MFCD00031395 | |
sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid | |
sodium;2-hydroxypropane-2-sulfonic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-761-2
RTECSNumber : TZ6550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only