Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-1-naphthoic acid, 98%

$248.62 - $248.62

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-1-naphthoic acid
CAS 2283-08-1
Molecular Formula C11H8O3
Formula Weight 188.18
MDL Number MFCD00004009
Beilstein 975575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1342630

  SDS
alfa aesar™
L1342630
250g Each for $248.62
Description & Specifications

Specifications

Formula Weight 188.18
Chemical Name or Material 2-Hydroxy-1-naphthoic acid
MDL Number MFCD00004009
Molecular Formula C11H8O3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 2283-08-1
Assay 98%
Beilstein 975575
Melting Point ≈167°C
TSCA TSCA
Quantity 250g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.